Would you like to log in again?
Pre-Cleaned Straight-Sided Tall Glass Jars, Clear, Environmental Express
Supplier: Environmental Express
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
APC1040 APC3120 APC3040 APC1130 APC1120 APC3130
23050-120CS 162.28 USD InStock
23050-120 23050-294 23050-150 23050-080 23050-128 23050-278
Pre-Cleaned Straight-Sided Tall Glass Jars, Clear, Environmental Express
Jars General Use Jars
These clear, straigt-sided glass jars come with polypropylene closure and PTFE liner.
All containers meet one of three levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance. Level I pre-cleaning is the same as Level II, but instead of a Statement of Compliance, a Certificate of Analysis is provided indicating that the containers received full EPA quality assurance treatment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty