Would you like to log in again?
ORION® Ultra-Micro Combination pH Electrode, Thermo Scientific
Supplier: Thermo Scientific
ORION®
9810BN
23227-006EA 1016.3 USD InStock
23227-006
ORION® Ultra-Micro Combination pH Electrode, Thermo Scientific
Electrodes pH Electrodes
Designed for long, thin test tubes.
Quick response time; requires only 0.5 µl of sample. Compatible with all Thermo Scientific pH and ISE/pH meters.
Ordering information: Comes complete with one bottle of reference filling solution 34108-020, adapter to fit a standard electrode holder, and filling pipet.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty