Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® 99M/59S Timer™ Timer
Supplier: VWR
VWR®
90013-34
23609-196EA 56.62 USD InStock
23609-196
VWR® 99M/59S Timer™ Timer
Timers Digital Timers
Features three-button control and large 1.9 cm (³/₄") digits readable from 6 m (20') away.
  • Traceable®
  • Countdown alarm
  • Magnet/stand/clip back
Timer counts down from 99 minutes, 59 seconds in one second increments. Memory function automatically returns to the last programmed time for repetitive tests. Alarm sounds for 30 seconds.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two-year AAA alkaline battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty