Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Traceable® Original Lab Timer
Supplier: VWR
VWR®
90012-84
23609-200EA 116.35 USD InStock
23609-200
VWR® Traceable® Original Lab Timer
Timers Digital Timers
Portable 3-channel timer signals drying and incubating times, helps monitor test results, and alarms for appointments.
  • Simple-to-use timer
  • Ideal for quality-control and experimental requirements
  • Alarm sounds for 60 seconds or may be turned off manually
  • Timing capacity to 9 hours, 59 minutes and 59 seconds
  • 1-second resolution
Timer is ideal for quality control and experimental requirements. Features 6.4 mm (¹/₄") high LCD digits. Programming capability for three different activities for 9 hours, 59 minutes, 59 seconds to 1 second, with 1-second resolution. Each channel has a distinct electronic alarm and display indicator. Alarm can be turned off manually or is silenced automatically after one minute. Unit is lightweight and portable, with chemical-resistant ABS housing.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two AA batteries.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty