Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® 60-Minute Interval Timer
Supplier: VWR
VWR®
90027-70
23609-218EA 42.16 USD InStock
23609-218
VWR® 60-Minute Interval Timer
Timers Mechanical Timers
This interval timer is adjustable from 0 to 60 minutes in one-minute intervals.
  • Keeps track of time in one-minute intervals
  • Easily audible bell alerts users to cycle end
Setting pointer to desired interval winds the time automatically. Indicating pointer always shows exact time remaining on easy-to-read numerals.
Ordering information: Supplied with a chemical-resistant case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty