Would you like to log in again?
Clay Annealing Cups, DFC Ceramics
Supplier: DFC Ceramics
C64020004 C64020002 C64020000
23982-049PK 96.9 USD InStock
23982-049 23982-027 23982-060
Clay Annealing Cups, DFC Ceramics
Crucibles
Cups are used for annealing gold buttons and for ignition of precipitates, such as barium sulfate, copper sulfide, silica, etc.
  • Smooth, unglazed surface
  • Supplied without cover
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty