Would you like to log in again?
Nunc® Storage Vials with Caps, Thermo Scientific
Supplier: Thermo Scientific
Nunc®
264300 264262
24270-128CS 329.34 USD InStock
24270-128 24270-126
Nunc® Storage Vials with Caps, Thermo Scientific
Vials
These vials with caps are designed for centrifugation and storage of PCR reagents, restriction enzymes and other diagnostics.
  • Made from a high density polypropylene copolymer
  • Excellent chemical resistance
The closures provided are high density polyethylene and leak proof.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty