Would you like to log in again?
Graduated Bottles, Dynalon
Supplier: Dynalon
202445-2000 202445-0050 202415-0050 202415-2000 202415-1000 202415-0100
24774-922CS 158.33 USD InStock
24774-922 24774-900 24774-912 24774-910 24774-908 24774-902
Graduated Bottles, Dynalon
Bottles Plastic Bottles
Economical graduated bottles are flexible, translucent, and have wide mouths for easy filling.
Maximum operating temperature for LDPE is 80 (176 °F) and 135 °C (275°F) for Polypropylene. A specially designed polypropylene closure provides a leakproof seal and has a hole for tagging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty