Would you like to log in again?
Tweezers, PTFE, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
316514-0000 316504-0001 316514-0001 316504-0002 316514-0002
24775-029CS 166.78 USD InStock
24775-029 24775-035 24775-025 24775-027 24775-033
Tweezers, PTFE, Dynalon
Forceps
Totally inert and non-contaminating forceps are manufactured from PTFE, making them suitable for use up to 260°C (500°F).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty