Would you like to log in again?
Nalgene® Economy Polypropylene Graduated Cylinders, Thermo Scientific
Supplier: Thermo Scientific
3664-0100 3664-1000 3664-0010 3664-0250 3664-0025 3664-0050 3664-0500
24780-265CS 123.51 USD InStock
24780-265 24780-287 24780-301 24780-367 24780-268 24780-323 24780-345
Nalgene® Economy Polypropylene Graduated Cylinders, Thermo Scientific
Cylinders Measuring Cylinders
Nalgene® Economy PP Graduated Cylinders exhibit excellent chemical resistance and have a generous pour spout for easily dispensing liquids
They have large rounded bases and are ribbed for reinforcement and stability.The cylinders are calibrated TC and TD at 20°C. Also they feature good temperature resistance up to 135°C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty