Would you like to log in again?
Spectra/Por® 3 Dialysis Membranes, MWCO 3500, Spectrum® Laboratories
Supplier: Spectrum
Spectra/Por®
132724 132725 132720 S632724
25219-121EA 697.05 USD InStock
25219-121 28170-166 25219-085 25219-041
Spectra/Por® 3 Dialysis Membranes, MWCO 3500, Spectrum® Laboratories
Tubing Dialysis Tubing
For dialysis and ultrafiltration in biochemical work.
Molecular weight cut-off is 3500. Pure, regenerated, natural cellulose. Free from fixed charges. Contain some glycerin and few impurities, which may be removed by washing. Porosity throughout roll is usually very uniform if roll is preserved carefully. Will change only slowly over a period of months if kept in the original polyethylene bag and refrigerated.
Roll length: 15.2 m (50')
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty