Would you like to log in again?
Spectra/Por® Universal Closures, Spectrum® Laboratories
Supplier: Spectrum
Spectra/Por®
142176 142154 142173 142170 142113 142252 142155 142112 142172 142253 142152 142174 142153 142156 142110 142175 142150 142250
25225-308PK 187.28 USD InStock
25225-308 25225-296 25225-310 25225-298 25225-320 25225-292 25225-294 25225-316 25225-304 25224-082 25225-318 25225-306 25224-080 25225-312 25225-300 25225-322 25225-314 25225-302
Spectra/Por® Universal Closures, Spectrum® Laboratories
Closures Test/Sample Tube Closures
Nylon closures lock gently over rigid membranes such as cellulose ester (CE) and polyvinylidene fluoride (type F) tubing, providing a strong, leakproof seal
Closures can also be used with traditional regenerated cellulose (RC) tubing. Sink in water. Can be sterilized with gamma radiation, ethylene oxide, ethanol, or formaldehyde. Not autoclavable. For best results, select a closure 4 to 10mm wider than the flat width of the membrane or tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty