Would you like to log in again?
Chemware® Teflon® Resin Evaporating Dishes, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Chemware®
D1069045 D1069044
25337-059EA 85.92 USD InStock
25337-059 25337-070
Chemware® Teflon® Resin Evaporating Dishes, Saint-Gobain Performance Plastics
Trays/Dishes/Pots Evaporating Basins/Dishes
With broad pouring spouts and flat bottoms
These resin evaporating dishes, saint-gobain performance plastics are made of PTFE.
Chemically inert to virtually all materials. Heat resistant to 260°C (550°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty