Would you like to log in again?
Chemware® PFA Evaporating Dishes, Saint-Gobain Performance
Supplier: Saint-Gobain
Chemware®
D1069543
25337-080EA 92.26 USD InStock
25337-080
Chemware® PFA Evaporating Dishes, Saint-Gobain Performance
Trays/Dishes/Pots Evaporating Basins/Dishes
The PFA Evaporating Dish design promotes evaporation of excess fluids to produce a concentrated solution or solid precipitate.
  • Chemically inert
  • Ideal for trace metal analysis
  • Transparent for easy viewing
  • Perfect for microwave use
  • Smooth, easy pouring due to broad spout
  • Capacity to brim: 116 mL
The flat bottom provides good surface contact with a heat source, while the non-stick angled walls direct fluids to the base, which limits condensation on the dish wall. This PFA dish can be used in a wide range of temperatures from cryogenics to 500 °F (260 °C). The dish is also ideal for holding solid samples or small volumes of liquid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty