Would you like to log in again?
Falcon® Tissue Culture Dishes, Polystyrene, Sterile, Corning
Supplier: Corning
Falcon®
353004 353025 353001 353003 353002 353003 353001 353002 353004 353025
25382-166CS 410.11 USD InStock
25382-166 25382-100 25382-188 CA25382-064 25382-064 CA25383-103 CABD353003 CA25382-188 25383-103 CA25382-100
Falcon® Tissue Culture Dishes, Polystyrene, Sterile, Corning
Culture Dishes Cell Culture Dishes
Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment.
Easy grip dishes offer a more secure hold when handling small dishes. The frosted handling rim of the dish base makes it easy to differentiate the base from the lid, allowing users to pick up the dish without accidentally removing the lid.
Dish 25383-103 features an injection-molded 20mm grid on the outside of the base to facilitate cell counting.
They are optically clear to allow for microscopic observation of cells. Stacking rings allow for easy handling and more stable stacking of dishes. Special venting features associated with the lid facilitate consistent gas exchange during incubation. Sterilized by gamma irradiation, nonpyrogenic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty