Would you like to log in again?
Corning® Culture Dishes, Nontreated, Sterile, Corning
Supplier: Corning
Corning®
430597 430591 431111 430589 431301 430588 431272
25382-452CS 248.28 USD InStock
25382-452 25382-450 25382-470 77776-922 29186-491 77776-742 25382-456
Corning® Culture Dishes, Nontreated, Sterile, Corning
Culture Dishes Cell Culture Dishes
Optically clear PS. For applications where cell attachment is not desired.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilized by gamma radiation
  • Certified non-pyrogenic
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty