Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Corning® Primaria™ Cell Culture Dishes, Corning
Supplier: Corning
Corning® Primaria™ Corning®
353801 353803 353802 353803 353801 353802
25382-701CS 773.91 USD InStock
CA25382-701 25382-701 CA25382-687 CA25382-654 25382-654 25382-687
Corning® Primaria™ Cell Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
PS, sterile, with lid.
  • Unique, nitrogen-containing tissue culture surface chemistry improves attachment, spreading and growth for many primary cells or cell lines
  • Optically clear and no special storage required
  • Convenient, peel open, medical-style packaging
The gases used to manufacture Corning® Primaria™ contain both oxygen and ammonia, resulting in the incorporation in the surface of a variety of nitrogen-containing functional groups in addition to the negatively charged oxygen containing groups found on traditional tissue culture treated surfaces. The incorporation of nitrogen containing cations has been correlated to attachment and spreading of primary endothelial cells in a clonal cell-growth assay. The complex surface on Corning® Primaria™ products is homogeneous and stable and is used to improve attachment and differentiation of a variety of cell types. The surface chemistry of Corning® Primaria™ products is confirmed by Electron Scanning for Chemical Analysis (ESCA).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty