Would you like to log in again?
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR
VWR®
25384-146 25384-144 15704-088 25384-148 15704-085 15704-087 15704-092 25384-078
15704-087CS 373.54 USD InStock
15704-087 15704-088 25384-146 25384-144 15704-092 25384-078 25384-148 15704-085
VWR® Specimen Containers, PP with PE Caps
Sample Containers General Purpose Containers
Durable, translucent containers may be used for storage or transport of most liquid or solid specimens.
  • Suitable for storage and transport of liquids or solids
  • Avoirdupois and metric unit graduations
  • Translucent
  • Polypropylene body with polyethylene caps
Containers tolerate strong acids, bases, and most organic solvents. Graduated in both avoirdupois and metric units. Withstand temperatures -10 to 130°C (14 to 266°F) continuously or -10 to 140°C (14 to 284°F) for a short period.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty