Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Disposable Petri Dishes
Supplier: VWR
VWR®
3505 3596 3506 3535 3501A 3561 3519 3305 3571WI 3527 3577 3515 3561WI 3502 3571 3511 3562 3552 3548NI 3517 3574 3508 3550 3516 3521 3306 3565 3504 3573 3503 3501 3509 3520 3525 3572 3310 3572WI 3308 3518
25384-318CS 553.52 USD InStock
25384-318 25384-312 89137-760 25384-310 25384-332 25384-338 25384-316 89038-970 25384-314 10799-192 25384-094 25384-093 10799-194 25384-092 89038-968 89022-320 25384-090 25384-308 25384-328 89230-472 25384-306 25384-322 25384-168 25384-344 25384-320 25384-342 25384-166 10053-704 25384-348 25384-304 25384-326 89107-632 25384-324 25384-302 25384-346 25384-062 25384-088 25384-164 10799-140
VWR® Disposable Petri Dishes
Culture Dishes Petri Dishes
Clearly superior, high-quality Petri dishes are manufactured from virgin crystal grade polystyrene for optimum flatness and high optical clarity.
  • Virgin crystal grade polystyrene construction
  • Available in varied sizes and styles for specialized and routine applications
  • Stacking ring and slip-ring features available
  • Supplied sterile
VWR® Petri dishes are available in a variety of sizes and styles to accommodate both specialized and routine needs. The stacking ring feature, on some models, facilitates positive stacking while the slip-ring feature, on other models, facilitates lateral or automated handling. Dishes are sterilized by irradiation or EO and are packaged in linear tear, medical style bags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty