Would you like to log in again?
Nalgene® Petri Dish Rack, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
5922-0024 5921-0060 5920-0060
25405-104CS 424.01 USD InStock
25405-104 25405-159 25405-170
Nalgene® Petri Dish Rack, Thermo Scientific
Racks Petri Dish Racks
Designed for holding plastic disposable petri dishes, these polycarbonate racks provide full side visibility for checking cultures during incubation
Lightweight, easy to load, safe for transporting. Used for media preparation, inoculation, incubation, and storage.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty