Would you like to log in again?
Tissue-Tek® Embedding Ring System, Sakura® Finetek
Supplier: Sakura
Tissue-Tek®
4151 4132 4114 4124 4115 4113 4133
25608-802CS 182.17 USD InStock
25608-802 25608-768 25608-800 25608-810 25608-766 25608-786 25608-764
Tissue-Tek® Embedding Ring System, Sakura® Finetek
Embedding Cassettes Embedding Rings
The Tissue-Tek® Embedding Ring System includes reinforced polystyrene embedding rings, which provide excellent support for clamping and sectioning, and reusable stainless steel base molds for specimen embedding
Rings come in four colors and fit all sizes of molds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty