Would you like to log in again?
Tissue-Tek® Mold Release Concentrate, Sakura® Finetek
Supplier: Sakura
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tissue-Tek®
4141
25608-804CS 235.38 USD InStock
25608-804
Tissue-Tek® Mold Release Concentrate, Sakura® Finetek
Moulds Moulds for Histology
For coating base molds to ensure clean separation of embedding paraffin from mold.
Dip molds in a solution of 5 mL release concentrate and 95 mL ethyl/isopropyl alcohol. Allow to dry prior to paraffin filling. Supplied in a 4 oz. bottle.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty