Would you like to log in again?
Tissue-Tek® Slide Staining Set, Sakura
Supplier: Sakura
Tissue-Tek®
4451
25608-902EA 950.96 USD InStock
25608-902
Tissue-Tek® Slide Staining Set, Sakura
Staining Boxes
Designed for histological or cytological staining
Provides simultaneous use of up to 12 solution wells, each deep enough to permit total immersion of 2.5x7.6cm (1x3") slides. Snap-locking cover on solvent-resistant metal frame prevents spills.
Ordering information: Complete set includes 10 white polypropylene dishes, 2 green solvent-resistant dishes, 12 lids, 12-unit metal carrier, and drain tray/cover. Additional staining dishes and 24-slide holder with detachable handle are sold separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty