Would you like to log in again?
Tenderfoot® Heel Incision Devices
Supplier: Accriva Diagnostics
Tenderfoot®
TFP50I
25718-040CS 1941.84 USD InStock
25718-040
Tenderfoot® Heel Incision Devices
Blood Lancets
These single-use heel-stick devices feature a highly sharpened surgical blade that swings across the skin in a gentle arc.
  • Sterile
  • Choice of devices suitable for preemies (low birth weight) or newborns (birth - six months)
The incision produced is through the rich capillary bed, but is carefully controlled to a depth that is superficial to most nerve fibers. The result is a high quality blood sample with significantly less pain and trauma to the skin.
Once the blade is triggered, it retracts permanently back into the device, thereby reducing the possibility of accidental sticks. The need for repeat sticks is virtually eliminated because each device is carefully controlled to produce an incision at a standardized length and depth.
Packaging: Each device is individually packaged.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty