Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 20–25, Non sterile, Aspen Surgical
Supplier: Aspen Surgical
Bard-Parker® Rib-Back™
371321 371320 371325 371322 371323
89176-378CS 162.52 USD InStock
89176-378 25860-042 25860-020 25860-064 25860-086
Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 20–25, Non sterile, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Blades
For use with No
4 and No. 6 Bard-Parker® scalpel handles. Fine blade carbon steel. Rib-Back™ is designed for greater cutting control and strength. Specially ground and carefully inspected. Blades individually packed in foil pouch.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty