Would you like to log in again?
VWR® FID Gas Stations
Supplier: VWR
VWR®
FID-2500-L1466 FID-1000-L1466
26000-036EA 28107.36 USD InStock
26000-036 26000-034
VWR® FID Gas Stations
Gas Generators Zero Air Generators
VWR® FID Gas Stations are designed to provide fuel gas and support air to flame ionization detectors, flame photometric detectors, and total hydrocarbon analyzers.
  • Eliminate dangerous gas cylinders from lab
  • Recommended and used by major GC and column manufacturers
VWR® FID Gas Stations provide both UHP zero air at purity levels below 0.05ppm total hydrocarbon content and hydrogen gas at purity levels above 99.9995% in one enclosure. Zero air is produced by purifying on-site compressed air. Pure hydrogen gas is generated by the electrolytic dissociation of water before passing through a desiccant cartridge. Systems increase the accuracy of analysis and reduce the need to clean detectors.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty