Would you like to log in again?
Barnstead™ Reverse Osmosis Distribution Pumps, Thermo Scientific
Supplier: Thermo Scientific
H1142
26139-720EA 18081.46 USD InStock
26139-720
Barnstead™ Reverse Osmosis Distribution Pumps, Thermo Scientific
Pumps Drum/Transfer Pumps
Depend on the heavy-duty, split-phase induction motor in Thermo Scientific™ Barnstead™ Reverse Osmosis Distribution Pumps.
  • Plastic pump bypass with restrictive orifice for tank
  • Low-level cutoff switch to shut pump off on storage tank when there are low water conditions
  • Heavy-duty, fully enclosed, split-phase induction motor
Distribution pumps are used with purified water storage tanks when it is necessary to keep the pure water under pressure for delivery to locations above tank level or at greater than gravity flow pressure.
Pump suction: 3/4"
Pump discharge: 1/2"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty