Would you like to log in again?
Barnstead Mega-Pure™ Glass Stills, Thermo Scientific
Supplier: Thermo Scientific
Mega-Pure™
A440266 A440267 A440696 A440518 A440697 A440367
26296-006EA 14740.86 USD InStock
26296-006 26296-003 26295-001 26294-257 26293-016 26293-017
Barnstead Mega-Pure™ Glass Stills, Thermo Scientific
Water Purification Systems Stills
Barnstead Mega-Pure Glass Stills effectively remove inorganic solids, organics, with boiling points higher than water, bacteria, and pyrogens
The stills are constructed of non-leaching components to ensure high purity water.
Certifications: CSA.
Ordering information: Automatic Collection System (ACS) is available for all Mega-Pure systems. The ACS automatically turns on/off the still to ensure a ready supply of distilled water at all times. The ACS comes standard with 45L glass storage bottle, float, and controller. For more information contact your Avantor representative.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: Thermo Scientific provides several service options for Thermo Barnstead Water System such as Installation, Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Barnstead Water System Technical Services" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty