Would you like to log in again?
Barnstead™ B-pure™ Filters, Thermo Scientific
Supplier: Thermo Scientific
B-pure™
FL583X1 FL583X4 FL583X6 FL583X5 FL583X2 FL583X3
26302-445EA 566.88 USD InStock
26302-445 26302-444 26302-441 26302-440 26302-443 26302-442
Barnstead™ B-pure™ Filters, Thermo Scientific
Filters Liquid Process Filters
Filters for pre- and post-filtration of water and are compatible with B-pure™ ½ size filter holders.
  • Features a quarter-turn quick-release canister for easy replacement
  • Natural polypropylene construction eliminates recontamination
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty