Would you like to log in again?
Barnstead B-pure™ Cartridge Water Purification System, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B-pure™
D4521 D4511 D4524 D4505
82002-642EA 2155.73 USD InStock
82002-642 26302-448 26302-250 26302-253
Barnstead B-pure™ Cartridge Water Purification System, Thermo Scientific
Water Purification Systems Deionizers
Thermo Scientific™ B-Pure Water Purification System is customizable for all pretreatment or deionization needs
Modular design provides the ability to add additional holders as needed. Quarter-turn, quick-release canisters make cartridge changes easy. All models accept Thermo Scientific Pura-Lite™ indicator to monitor water quality.
Pura-Lite™ indicators are easy-to-read lights. Green light indicates acceptable water purity, and red light indicates unacceptable water purity. Cell thread is 12.7 mm (1/2") NPTM.
B-Pure water purification system is an economical, laboratory-grade water system that can be customized to meet specific application needs. Recommended applications include pretreatment and deionization.
Ordering information: Canisters sold separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty