Would you like to log in again?
PYREX® Rotary Evaporator Trap, with [ST] Inner and Outer Joints, Corning
Supplier: Corning
PYREX®
4000-10024 4000-25024 4000-10019
26307-264CS 262.69 USD InStock
26307-264 26307-262 26307-270
PYREX® Rotary Evaporator Trap, with [ST] Inner and Outer Joints, Corning
Vacuum Traps
These PYREX® traps are used in rotary evaporators. They have a round body with upper and lower stems.
  • Lower stem has a Standard Taper inner joint at the bottom and extends upward inside the body
  • The closed top of this stem has two side openings
  • The upper stem has a Standard Taper outer joint
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty