Would you like to log in again?
WHEATON® Crimpers and Decappers, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
224316 224357 224323 224322 224307 224321
10816-282CS 1670.75 USD InStock
10816-282 26676-406 14217-402 26676-426 14217-400 26676-413
WHEATON® Crimpers and Decappers, DWK Life Sciences
Crimpers and Decappers
These crimpers are used for flip cap seals and for aluminum seals sizes 16.5, 28.5, and 30 mm.
  • 1 year warranty
  • Not autoclavable
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty