Would you like to log in again?
CONDUCTIVE RBR24X36 BLK TYPE I
Supplier: DESCO
Conductive Antifatigue Mat^ Designed to inerlock and create a covering for any size or shape. Grease and chemical resistant. Features a bubble dome shape, making it a softer, premium mat for antifatigue. Type i "interlocking" mat. Provides conductive ESD protection. Black. 1.3cm thick. 61x91cm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONDUCTIVE RBR24X36 BLK TYPE I 26911-922 40930 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty