Would you like to log in again?
Electro-Analyzer, Model E1000, Eberbach
Supplier: Eberbach
E1000.00
27190-009EA 18937.97 USD InStock
27190-009
Electro-Analyzer, Model E1000, Eberbach
Analyzers Elemental Analysers
Eberbach's Electro-Analyzer E1000 apparatus is used for the determinations of lead, nickel, copper, zinc, antimony, cadmium and other metals by the electro-deposition process
Unit has a finished steel case (30" high) with stainless steel front surface and occupies a table space of 15" x 17". The front panel protects the apparatus from corrosive spillage and fumes. No electrically corrodible components are positioned where material may fall into the cell and alter results. The unit delivers up to 10 volts, 10 amperes DC at each of two positions for simultaneous determinations. Separate ammeters (0-10A) and center-zero voltmeters (10-0-10V) indicate amount and direction of current flow at each position. Polarity reversing switches change the polarity of the electrodes. Current to each position is supplied by individual power transistors controlled by potentiometers. In line fuses protect AC and DC circuits. Stirrers are driven at a constant speed of 620 rpm by a split-phase induction motor. Hollow spindles permit easy vertical adjustment of the glass stirring rods. Power to each stirrer and entire unit is controlled by individual switches with pilot lamps. Heavy-duty platinum electrodes (45 mesh) are recommended for this unit however, lighter weight electrodes may be used. Spring loaded electrode holders grip the electrodes without deforming the stem and may be moved horizontally for centering nested electrodes. For rotating anode techniques a stainless steel rotating anode holder may be substituted for glass stirring rod.
Ordering information: Manufacturer's two-year parts and labor warranty. Unit ships with 2 E1060 glass agitators. Eberbach Platinum Anode (E1100) and Platinum Cathode (E1120) available; contact Avantor for further information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty