Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Glass Plates for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Supplier: Thermo Scientific
Owl™
P1-14R P10-20R P5-16R ADJ1-40R ADJ1-38G ADJ1-23G P2-20FR P2-20FG P1-14G ADJ1-40G P1-12G
27372-014EA 34.36 USD InStock
27372-014 27373-309 27373-441 27372-580 27372-020 27372-582 27372-516 27373-428 27372-522 27372-016 27372-524
Glass Plates for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Electrophoresis Plates and Sheets
Plates are available in three styles: notched glass, blank glass, and offset glass.
Heat sinks are recommended when running sequencing gels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty