Would you like to log in again?
Combs for Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1, B2 and B3, Thermo Scientific
Supplier: Thermo Scientific
EasyCast™
B1-5C B2-16 B1-5D B1-10 B2-WALL B1A-5D B1A-10 B1-8C B1-12D B2-RL-25D B1A-8 B1-PREP B2-24 B2-RL-25C B1-14 B1A-6 B2-20 B1A-PREP B1A-12 B2-8D B1-RL-17D B1A-5C B1-12C B2-RL-9C B1-8D B1-RL-9C B2-12 B2-PREP B2-RL-9D B1-RL-17C B2-8C B1-RL-9D
27372-222EA 95.16 USD InStock
27372-222 27372-288 14006-030 14005-896 27373-284 14005-894 27373-282 14005-898 14007-028 14005-892 27372-224 27373-280 14005-890 27373-313 27373-314 27372-290 27373-278 27373-312 27373-317 27373-318 27373-315 27373-316 27373-272 14005-886 14005-884 27373-276 27373-254 27373-274 14005-888 14005-882 27372-214 27372-216
Combs for Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1, B2 and B3, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Use comb with your exisitng U.V
  • For use in Owl™ EasyCast™ Mini Gel Electrophoresis Systems
  • Cast smaller gels
For use in Owl™ EasyCast™ Mini Gel Electrophoresis Systems. Use comb with your exisitng U.V. Transmissible (UVT) gel tray and comb slots to cast smaller gels. Thickness: 1.5mm (1/16").
Transmissible (UVT) gel tray and comb slots to cast smaller gels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty