Would you like to log in again?
Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1806-WF 1806-WF 1806-WFB
89176-730CS 395.6 USD InStock
89176-730 CA27500-100 27500-100
Puritan® Mini-Tip Foam Tipped Applicator, Wooden Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
These swabs have thermal edge seals and thermal bonding to the nadle help ensure the sponge-like foam will clean up unwanted particles without releasing any into your critical environment.
Made in the USA.
Certifications: CE marked.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty