Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
WHEATON® Replacement Silicone Stoppers for Filtration Assemblies, DWK Life Sciences
Supplier: DWK Life Sciences
419356 419336
27589-108CS 153.1 USD InStock
27589-108 27589-092
WHEATON® Replacement Silicone Stoppers for Filtration Assemblies, DWK Life Sciences
Stoppers Silicone Stoppers
This unit is designed for filtering viscous or proteinaceous solutions or to produce ultraclean filtrate.
These filtration assemblies are recommended for routine filtration of aqueous solutions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty