Would you like to log in again?
WHEATON® Filtration Assembly with Stainless Steel Support and No. 8 Stopper Connections, 47 mm, DWK Life Sciences
Supplier: DWK Life Sciences
419360
27589-110CS 464.8 USD InStock
27589-110
WHEATON® Filtration Assembly with Stainless Steel Support and No. 8 Stopper Connections, 47 mm, DWK Life Sciences
Funnels Filtration Funnels Filtration Apparatus Glass
The filtration assembly with fritted glass support is designed to handle up to 1000 ml of liquid for analysis of particulate or microbiological contamination.
  • Glass components are manufactured from WHEATON 33 low extractable borosilicate glass
  • Recommended for filtering viscous or proteinaceous solutions or to produce ultraclean filtrate
Flasks have a #2 hose connection for 1/4” (6 mm) I.D. tubing. Flasks are not included.
Certifications: Glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Delivery information: Includes a 316 stainless steel support with 120 mesh screen, PTFE support screen gasket, glass support base, 300 ml graduated funnel, anodized aluminum clamp and a No. 8 silicone stopper.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty