Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Cellulose Soxhlet Extraction Thimble, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whatman™
2810-166 2810-339 2810-166 2800-300 2800-373 2800-250 2800-308 2810-338 2800-266 2800-288 2800-280 2800-331 2800-105 2800-258 2800-199 2800-280 2800-412 2800-259 2800-300 2810-266 2800-259 2800-199 2800-226 2800-432 2800-228 2810-432 2800-308 2800-185 2810-228 2810-228 2800-330 2800-258 2800-330 2800-608 2800-331 2810-266 2800-185 2800-288 2800-282 2800-412 2810-338 2800-339 2800-282 2800-432 2800-105 2800-338 2800-250 2800-338 2800-266 2810-432 2800-226 2800-260 2800-373 2800-339 2810-339 2800-608 2810-902 2810-258 2800-228 2800-260 2810-258
27732-065PK 244.32 USD InStock
27732-065 CA11013-710 CA27730-142 CA27730-183 27732-145 CA27730-140 CA28297-430 CA28297-432 CA27730-106 CA28297-436 CA27730-026 76332-602 CA11012-824 CA27730-060 CA11013-722 27730-048 CA11013-720 CA27732-087 CA27732-167 CA27732-123 27730-128 CA70250-016 27730-084 27730-161 27730-081 27732-087 CA27730-084 CA27730-161 27732-123 27732-167 27730-117 CA27730-128 CA11012-856 CA27730-048 10035-394 70250-016 10035-396 10035-390 10035-392 CA70250-022 CA27730-081 10035-398 28297-436 27730-026 27730-142 28297-432 CA27732-065 CA11012-860 CA27732-145 27730-106 CA27730-117 CA11012-864 CA11012-858 10035-402 70250-022 27730-060 28297-430 10035-404 27730-140 27730-183 10035-400
Whatman™ Cellulose Soxhlet Extraction Thimble, Whatman products (Cytiva)
Extractors Extraction Thimbles
Cytiva's Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems

  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1 to 1.5 mm to support application need
  • Option for double thickness for added rigidity

Standard extraction thimbles have a wall thickness of 1 to 1.5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.

Used in air and waste gas analysis.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty