Would you like to log in again?
Whatman™ TCLP Testing Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1810-150 1810-125 5925-142 1810-142 1810-125 1810-142 1810-047 1810-090 1810-090 5925-142 1810-047 1810-150 1810-110 1810-110
28137-662PK 137.22 USD InStock
CA28137-660 28137-662 CA28137-662 CA28297-308 28137-664 28137-670 28137-660 CA28137-670 CA28137-668 28137-666 CA28137-664 CA28137-666 28137-668 28297-308
Whatman™ TCLP Testing Filters, Whatman products (Cytiva)
Filters Glass Fiber Filters
Cytiva's Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
Toxicity Characteristic Leaching Procedure (TCLP) is designed to predict the toxicity of hazardous waste. Industrial waste must comply with established TCLP limits to ensure that hazardous contaminants, known as landfill leachate, do not migrate into groundwater and threaten local drinking water sources.
By accurately characterizing the degree of contamination, material can be disposed of as safely and economically as possible.
To dispose of hazardous waste in a United States municipal landfill, TCLP limits must fall within standard levels set for maximum contamination levels (MCLs). If MCLs are exceeded, the producer must dispose of the materials at a specialized hazardous waste facility, which increases costs significantly.
Acid-treated, low metal Whatman TCLP test filters from Cytiva's business can help to determine what steps, if any, must be taken to mitigate landfill leachate risk.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty