Would you like to log in again?
Whatman™ Polydisc HD In-Line Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Polydisc™ Polydisc™ Whatman™
2228 6728-5050 6728-5100 6728-5100 2228 2227 6728-5050 2227
10035-760PK 635.55 USD InStock
10035-760 10035-758 10147-996 CA11013-066 28137-747 CA28137-747 CA10147-996 CA11012-820
Whatman™ Polydisc HD In-Line Filters, Whatman products (Cytiva)
Filters In-Line Filters
Polydisc HD (Heavy Duty) is a durable all-polypropylene in-line filter for large volumes of liquid samples containing even harsh chemical solvents.
  • In-line, hose-connected filter with up to 1000 ml capacity
  • High flow filter with durable all-polypropylene construction
  • Broad aqueous and solvent compatibility
  • 99% liquid retention rating
  • Available in 5 and 10 µm pore size
  • Recyclable after use
Labs that regularly process large sample volumes with different chemical compositions, pH values, and temperatures require a filter that is fast, durable, and broadly compatible.
Heavy-duty Polydisc filter is a durable all-polypropylene filter. Its high strength, aqueous and solvent compatibility, and flow rate make it an excellent in-line filter for up to 1000 ml sample volumes
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty