Would you like to log in again?
Whatman™ Polydisc™ TF In-Line Filters
Supplier: Whatman products (Cytiva)
Whatman™ Polydisc™ Polydisc™
6721-5010 6720-5002 6721-5010 6720-5001 6720-5002 6720-5045 6720-5001 6720-5045
28137-850PK 214.7 USD InStock
28137-850 28137-852 CA28137-850 CA28137-856 CA28137-852 28137-854 28137-856 CA28137-854
Whatman™ Polydisc™ TF In-Line Filters
Filters In-Line Filters
Whatman Polydisc TF In-Line Filters from Cytiva are lightweight devices with high chemical and solvent resistance that are particulraly suitable for protective vents or in-line filtration applications.
  • Chemically resistant hydrophobic PTFE membrane is well suited as a solvent filter for many different solutions
  • Filter can be autoclaved multiple times without losing integrity
  • Integrity-testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight to prevent tube collapsing
Filtering organic solvents and aggressive solutions calls for a chemically resistant, durable membrane. Cytiva's Polydisc TF features a PTFE membrane resistant to such compounds. The 1 µm membrane features a polypropylene prefilter for solvent filtration containing high particulate density in order to extend the useful life of the membrane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty