Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ VACU-GUARD™ 150 Capsule Filters, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™ VACU-GUARD™ Whatman™
6722-1002 6722-1002 6722-1001 6722-1001
28137-857PK 237.5 USD InStock
CA28137-859 28137-857 28137-859 CA28137-857
Whatman™ VACU-GUARD™ 150 Capsule Filters, Whatman products (Cytiva)
Filters In-Line Filters
Whatman VACU-GUARD capsule filters are ideal for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.

  • Excellent for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols
  • Designed for in-line use with stepped barb connections for 10 to 12 mm ID hose
  • Available with choice of chemical trap: Activated carbon, molecular sieve, or desiccant
  • Membrane retains 99.99% of airbone particles greater than 0.1 µm
  • Features hydrophobic polytetrafluoroethylene (PTFE) membrane

The easy-to-use VACU-GUARD inline filter devices help to confine and isolate infectious materials in vacuum systems and protect your laboratory.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty