Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ VACU-GUARD Disk Filters, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whatman™ VACU-GUARD™
6722-5001 6722-5001 6722-5000 6722-5000
28137-737PK 246.05 USD InStock
28137-737 28137-858 CA28137-737 CA28137-858
Whatman™ VACU-GUARD Disk Filters, Whatman products (Cytiva)
Filters In-Line Filters
Whatman VACU-GUARD disk filters are ideal for protecting vacuum pump systems from aqueous aerosols and water intrusion.

  • Designed for in-line use with stepped barb connections
  • Membrane retains 99.99% of airbone particles greater than 0.1 µm
  • Features hydrophobic polytetrafluoroethylene (PTFE) membrane

Available in 50 and 60 mm formats.

Ordering information: All materials pass USP Class VI test for plastics.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty