Would you like to log in again?
Whatman™ Polydisc AS In-Line Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™ Polydisc™
6724-5045 6724-5002 6724-5145 6724-5002 6724-5145 6724-5045
28137-476PK 697.3 USD InStock
28137-476 28137-862 CA28137-476 28137-864 CA28137-862 CA28137-864
Whatman™ Polydisc AS In-Line Filters, Whatman products (Cytiva)
Filters In-Line Filters
Whatman Polydisc AS (Aqueous Solution) in-line filters from Cytiva feature high-throughput PES membrane with a glass microfiber prefilter. Polydisc comes presterilized in medical-grade blister packs
  • Whatman filters with polyethersulfone membranes have low protein binding and are surfactant-free
  • GMF prefilter removes heavy particulates, increasing membrane longevity
  • Radiation sterilized, so there are no ethylene oxide (EtO) residuals
  • Barbed hose connections fit multiple tubing sizes
  • Integrity-testable with the bubble point method
  • Lightweight design prevents tubing collapse
  • Recyclable after use
AS Polydisc filters use polyethersulfone membranes, which are low protein binding and surfactant free, making them the recommended membrane for many pharmaceutical applications.
These membranes have been demonstrated to contribute low levels of extractables which might otherwise interfere with sample analysis. Polydisc AS also incorporates a glass microfiber prefilter to handle heavy particulates and extend membrane life.
Nonsterile filters can be sterilized in the lab by autoclaving, but filters must be tested afterwards to ensure integrity. AS Polydisc filters eliminate the need for autoclaving, because each filter is provided irradiated and presterilized in its own "medical grade" clear blister pack, secured in a protective shelf pack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty