Would you like to log in again?
Whatman™ HEPA-Cap Vent Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ HEPA-CAP™
6702-3600 2709T 6702-7500 6702-7500 2609T 6702-9500 6702-3600 2609T 6702-9500
28137-884EA 88.3 USD InStock
28137-884 11008-594 28137-886 CA11008-594 CA28137-884 CA14230-540 CA28137-886 14230-540 11008-598
Whatman™ HEPA-Cap Vent Filters, Whatman products (Cytiva)
Filters Venting Filters
High-retention and high flow-rate glass fiber filters for research and industrial use to remove particulates from air and gases.
  • Typical retention efficiency: 99.97% of all particles ≥0.3 µm in air
  • Depth filter design: Allows for high loading capacity and bi-directional flow
  • Repeatedly autoclavable: At 121 °C for 20 minutes (132 °C max.)
Whatman HEPA-CAP Vent Filters are used across scientific, research, and industrial environments in a variety of air and gas filtration applications that require high particle retention and the ability to capture dirt and debris at high flow rates.
In-line filter capsules are particularly suitable for sterile venting of fermenters or reaction vessels to prevent airborne particles from entering or exiting the vessel. They are also well suited for tissue culture, in-line filtration, and prefiltration for suction or to serve gas in-line filters.
The Whatman HEPA-CAP Vent Filter glass fiber filter media is strengthened by dual lamination with a tough polyester monofilament. The filters are pleated to maximize surface area for high particle retention and high flow rate. The filters are available in a range of different sizes, formats and end fittings.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty