Would you like to log in again?
Whatman™ Puradisc Syringe Filters, PP, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Puradisc™ Puradisc™
6788-1302 6790-2502 6790-2502 6788-2504 6786-2502 6785-1304 6785-1302 6788-2502 6785-1302 6786-2504 6788-1302 6788-2504 6788-1304 6788-1304 6785-1304 6790-2504 6790-2504 6786-2502 6788-2502 6786-2504
28137-960PK 781.85 USD InStock
28137-960 28137-542 CA28137-542 CA28137-960 28137-544 CA28137-954 CA28137-536 28137-956 28137-967 CA28137-967 28137-958 CA28137-956 28137-974 CA28137-544 CA28137-974 28137-536 28137-954 CA28137-958 CA10147-998 10147-998
Whatman™ Puradisc Syringe Filters, PP, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc micron syringe filters from Cytiva's offer a versatile solution for analytical sample preparation needs. They are a variety of options available to suit all labs needs, and are well-suited for routine syringe filtration of samples up to 100 ml for a range of applications, including high performance liquid chromatography (HPLC) and capillary electrophoresis (CE).
  • Choice of 13 or 25 mm Ø non sterile syringe filter to suit filtration sample volume
  • Sub-micron pore sizes, 0.2 and 0.45 μm, for fine particle retention to protect instruments and chromagraphy columns
  • Autoclavable for sterile sample preparation
  • Also available with depth polypropylene (DpPP) filtration medium
  • Recyclable after use
Whatman polypropylene syringe filters have high and uniform tolerance to heat and mechanical stress. The slightly hydrophobic membrane is resistant to a wide variety of organic solvents.
Cytiva's Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of filtration applications. Puradisc is available in a range of filtration media to filter samples of diverse chemical solvents, compounds and sample types. Filtration materials include: regenerated cellulose (RC), cellulose nitrate, nylon, polytetrafluoroethylene (PTFE), polyethersulfone (PES), polypropylene, polyvinylidene difluoride (PVDF), and cellulose fiber acetate.
Puradisc syringe filters are well-suited for a wide variety of sample types and applications, including capillary electrophoresis(CE), biological sample preparations and analysis, protein filtration, UV spectroscopy and high performance liquid chromatography (HPLC).
Cytiva also offers high-performance GD/X syringe filters with a multi-layer glass fiber prefilter for filtering larger volumes of harder-to-filter samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty