Would you like to log in again?
Whatman™ Aqueous IFD, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
6726-5002A 6726-5002 6726-5002A 6726-5002
80089-524PK 260.3 USD InStock
80089-524 28137-970 CA80089-524 CA28137-970
Whatman™ Aqueous IFD, Whatman products (Cytiva)
Filters In-Line Filters
Aqueous IFD from Cytiva is a polypropylene housed in-line filter/degasser (IFD) with nylon membrane. It connects to an HPLC line and simultaneously filters and degasses during HPLC mobile phase preparation.
  • Faster than traditional methods of HPLC mobile phase preparation, saving time in the laboratory
  • Provides pure filtration of aqueous mobile phases without expensive degassing equipment
  • Chemically resistant polypropylene construction enables reliable filtration
  • Can be integrity-tested with the bubble point method
  • Includes compression end fittings
  • Recyclable after use
The Aqueous IFD is designed to work as an in-line filter with mobile phases containing at least 20% of the aqueous component. It has a 0.2 µm hydrophilic nylon membrane to support aqueous-based HPLC mobile phase preparation.
Whatman Aqueous IFD have a polypropylene housing sealed by a security ring, fittings to accommodate 1/16”-1/8” tubing, and an air vent on the inlet with luer lock cap to enable priming.
An in-line filter works on the principle of "bubble point" – the point of pressure at which gases will pass through a wet membrane. If pressure is maintained below the bubble point, the gas will not pass through the membrane and is trapped by the filter device.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty