Would you like to log in again?
Whatman™ Anotop Plus Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™ Anotop™
6809-3002 6809-4002 6809-4022 6809-3012 6809-3012 6809-4024 6809-3022 6809-3022 6809-4012 6809-3002 6809-4024 6809-4022 6809-4002 6809-4012
28138-043PK 845.5 USD InStock
CA28138-041 28138-043 CA28138-031 28138-041 28138-031 CA28138-045 CA28138-043 28138-045 28138-029 CA28138-027 CA28138-039 28138-027 28138-039 CA28138-029
Whatman™ Anotop Plus Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Anotop Plus non-sterile syringe filters from Cytiva have an integrated glass microfiber prefilter and inorganic membrane, enabling filtration of a wide variety of sample solvents and compounds.
  • Capillary pore structure for well-defined particle filtration
  • Low protein binding filter for applications requiring high protein recovery
  • Integrated glass microfiber prefilter for filtering larger volumes of difficult-to-filter samples
  • Recyclable after use
High solid content samples can be difficult to filter because of their tendency to prematurely clog a single membrane filter. Each Anotop Plus syringe filter includes a glass prefilter designed to separate difficult, hard-to-filter samples without adversely affecting the filtration efficiency of the final membrane.
Inclusion of a prefilter can eliminate the need for expensive and time-consuming sequential filtration. Whatman Anotop Plus filters can be used with most organic solvents and aqueous materials and are suitable for sample volumes up to 100 ml. They come in two diameters and three pore sizes.
Common applications for Anotop Plus syringe filters include sample preparation prior to capillary electrophoresis or HPLC/UHPLC, as well as filtration of nanoparticles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty